Documentos clave
9025
CELLTREAT
VistaLab Pipette Tips
250µL Pipette Tips, MLA, Stacked Dense Rack, Non-sterile
Sinónimos:
pipette tips
About This Item
PDF Icon
Shipped & Sold By CELLTREAT
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
CELLTREAT
General description
A superior fit for your MLA & Ovation Pipettes. Use genuine VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation and MLA pipettes. All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time.
Stacked Rack
  • Non-sterile only
  • Tips and racks are manufactured from 100% virgin polypropylene and can be autoclaved
  • 200 tips/layer
  • For Small volume (250 mL) tips - 5 layers - 1000 tips
  • For Large volume (1250 mL) tips - 3 layers - 600 tips