MilliporeSigma
1537971002
Blickman
Blickman Paul Cabinet
47 5/8"W x 79 1/4"H X 16"D w/(5) S/S Shelves w/(3") Casters (2) w/Brake
About This Item
PDF Icon
Shipped & Sold By Blickman
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
Blickman
General description
Blickman's Paul cabinet is 79-1/4” tall on casters and 16” deep and includes five shelves adjustable every half inch. This cabinet provides maximum shelving storage for instruments and supplies. View its contents through its two glass doors.