Documentos clave
Z557765
Sigma-Aldrich
Aldrich® swivel McLeod vacuum gauge
replacement parts kit
About This Item
Código UNSPSC:
41112406
NACRES:
NB.79
Descripción general
Contains a set replacement o-ring seals and plastic nuts for the swiveling connection that joins the two glass components.
Made with permanent linear and square root ceramic scales on a white background, high-vacuum O-ring seals, precision bore capillaries, Delrin® plastic and black anodized aluminum parts. Accurate to ±3% of reading or 1mm of scale division, whichever is greater. Gauges include operating instructions but do not include oil or mercury fill. Swivel arrangement permits rigid connection to the vacuum system with the scales vertical at all times.
Información legal
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Delrin is a registered trademark of E. I. du Pont de Nemours and Company
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)