Key Documents
Z517607
Sigma-Aldrich
Aldrich® round-bottom storage flask with stopcock-equipped septum-inlet
capacity 1,000 mL, with 4 mm glass stopcock, joint: ST/NS 29/32
Sinónimos:
bottom flask, flask round, round flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
round bottom flask
single-neck
joint
joint: ST/NS29/32
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
1,000 mL
General description
Mit NS Schliff. Passende Septen: Z10,072-2 oder Z12,435-4 (siehe Abschnitt Septen).
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)