Documentos clave
TEM-76043-22
Sigma-Aldrich
Cryo-SiN TEM Windows
20 nm Thick, window size 500 μm , Single
About This Item
Código UNSPSC:
41111761
ventana tamaño
500 μm , Single
Descripción general
Silicon Nitride 20 nm thick TEM Windows - Single 500 micron window. Ideal substrate for Cryo-Microscopy. Avoid substrate degradation. Plasma cleanable to reduce contaminants. Large 500 micron windows provid 50x50 micron viewing from any orientation at 70 degreee of tilt for tomography.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)