MilliporeSigma
R549711
ETHYL (2-{[({5-[(2-{[4-(2-ETHOXY-2-OXOETHYL)-1,3-THIAZOL-2-YL]AMINO}-2-OXOETHYL)SULFANYL]-1,3,4-THIADIAZOL-2-YL}SULFANYL)ACETYL]AMINO}-1,3-THIAZOL-4-YL)ACETATE
AldrichCPR
Seleccione un Tamaño
50MG
$30.70
$30.70
Normalmente se envía en 2 días laborables. (Para pedidos fuera de Estados Unidos, calcule la entrega 1 o 2 semanas más tarde)
Seleccione un Tamaño
Cambiar Vistas
50MG
$30.70
About This Item
Fórmula lineal:
C20H22N6O6S5
Número de CAS:
Peso molecular:
602.755
Número MDL:
$30.70
Normalmente se envía en 2 días laborables. (Para pedidos fuera de Estados Unidos, calcule la entrega 1 o 2 semanas más tarde)
Otras notas
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)