Documentos
MT51343205
Mettler Toledo InLab® Redox Ag ORP electrode
Redox Electrode with Silver Ring Indicator and potassium nitrate Electrolyte
About This Item
UNSPSC Code:
39121436
NACRES:
NB.79
material
glass shaft
description
S7 connection
Without cable
manufacturer/tradename
Mettler-Toledo 51343205
temperature measuring range
0-100 °C
L × W
120 mm × 12 mm
General description
Mettler Toledo InLab® Redox Ag is a combination electrode for measuring the oxidation/reduction power of a solution. Consists of a silver ring indicator electrode and a silver/silver chloride reference electrode.
The measuring signal is produced on the surface of the precious metal by an exchange of electrons with the oxidation-reduction system of the measured medium.
Combination design avoids use of separate reference and indicating electrodes.
The innovative Safelock closing ring gives easy access for re-filling reference electrolyte and complete sealing when closed.
Different cables can be connected to the S7 screw head allowing use with a wide range of meters.
  • Combination design
  • Safelock closing ring
  • Detachable cable
Legal Information
InLab is a registered trademark of Mettler-Toledo, Inc.