Saltar al contenido
Merck

GF96610833

Nickel

foil, 8mm disks, thickness 0.3mm, annealed, 99%

Sinónimos:

Nickel, NI000562

Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización

Seleccione un Tamaño


Seleccione un Tamaño

Cambiar Vistas

About This Item

Fórmula empírica (notación de Hill):
Ni
Número de CAS:
Peso molecular:
58.69
Número MDL:
Código UNSPSC:
12141729
ID de la sustancia en PubChem:
NACRES:
NA.23

Ensayo

≥99%

Formulario

foil

fabricante / nombre comercial

Goodfellow 966-108-33

resistividad

6.97 μΩ-cm, 20°C

bp

2732 °C (lit.)

mp

1453 °C (lit.)

densidad

8.9 g/mL at 25 °C (lit.)

cadena SMILES

[Ni]

InChI

1S/Ni

Clave InChI

PXHVJJICTQNCMI-UHFFFAOYSA-N

Descripción general

For updated SDS information please visit www.goodfellow.com.

Información legal

Product of Goodfellow

Pictogramas

Health hazardExclamation mark

Palabra de señalización

Danger

Frases de peligro

Clasificaciones de peligro

Carc. 2 - Skin Sens. 1 - STOT RE 1

Código de clase de almacenamiento

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

Clase de riesgo para el agua (WGK)

WGK 1

Punto de inflamabilidad (°F)

Not applicable

Punto de inflamabilidad (°C)

Not applicable


Elija entre una de las versiones más recientes:

Certificados de análisis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documentos section.

Si necesita más asistencia, póngase en contacto con Atención al cliente

¿Ya tiene este producto?

Encuentre la documentación para los productos que ha comprado recientemente en la Biblioteca de documentos.

Visite la Librería de documentos

G E van Dorssen et al.
Journal of X-ray science and technology, 3(2), 109-117 (1992-01-01)
The structure of Ni-C multilayer and single nickel layer samples has been analyzed before and after annealing, using two techniques: fluorescence EXAFS (F1EXAFS) at the Ni-K. edge and CuKα reflection. Annealing at a temperature of 450°C resulted in a change
R J Schulz et al.
Physics in medicine and biology, 35(11), 1563-1574 (1990-11-01)
The thermal defects of A-150 plastic and graphite referenced to aluminium were determined for 800 keV protons scattered by a 2 micron nickel foil (mean transmitted energy = 550 keV). Composite cores of Al/A-150, graphite/A-150 and Al/graphite which could be
F E Cecil et al.
The Review of scientific instruments, 81(10), 10D326-10D326 (2010-11-09)
We have examined the observed currents in the front foils of the JET Faraday cup lost alpha particle diagnostic KA-2. In particular, we have sought to understand the currents during Ohmic plasmas for which the ion flux at the detectors
W K Winnett et al.
Talanta, 41(10), 1627-1630 (1994-10-01)
A modification of the sample introduction system for combustion total organic carbon analyzers is presented where sample is sprayed into the instrument. Thermally created sprays and nebulizer sprays are discussed. It is shown that spray introduction gives more symmetric signals
Alex Ignatiev et al.
Dalton transactions (Cambridge, England : 2003), (40)(40), 5501-5506 (2008-12-17)
Nanostructured thin film solid oxide fuel cells (SOFC) have been developed for reduced temperature operation, with high power density, and to be self reforming. A thin film electrolyte (1-2 microm thickness), e.g., yttria-stabilized zirconia (YSZ), is deposited on a nickel

Nuestro equipo de científicos tiene experiencia en todas las áreas de investigación: Ciencias de la vida, Ciencia de los materiales, Síntesis química, Cromatografía, Analítica y muchas otras.

Póngase en contacto con el Servicio técnico