Documentos clave
BR705384
Individual stand for pipette Transferpette® electronic + -8-12 electronic
Seleccione un Tamaño
1 EACH
$245.00
$245.00
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
1 EACH
$245.00
About This Item
Código UNSPSC:
41121508
NACRES:
NB.01
$245.00
Check Cart for Availability
Materiales
PBT
PC
envase
pack of 1 ea
fabricante / nombre comercial
BRAND 705384
Categorías relacionadas
Descripción general
Save space in the laboratory and use stands and wall mounts to safely store your BRAND single or multichannel pipettes. BRAND has the right solution for every model and every need:
Wall/rack mounts are available for mechanical single- and multichannel Transferpette® S pipettes. The single stand for the Transferpette® S mechanical pipette is usually used for the multichannel pipette. Single stands are also available for the Transferpette® electronic pipette - one stand for pipettes up to 1000 µl and another for the 5 mL pipette. The benchtop rack (carousel) stand holds up to six Transferpette® S or Transferpette® S -8/-12 pipettes, saving considerable space. Three Transferpette® electronic pipettes (single channel, up to 1000 μl) can be charged simultaneously in the charging stand. Individual power supplies no longer needed!
Información legal
Transferpette is a registered trademark of BRAND GMBH + CO KG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)