MilliporeSigma
224871
Sigma-Aldrich
Sodium bromate
≥99%
About This Item
Fórmula lineal:
NaBrO3
Número de CAS:
Peso molecular:
150.89
Número CE:
Número MDL:
Código UNSPSC:
12352300
eCl@ss:
38050507
ID de la sustancia en PubChem:
Ensayo:
≥99%
Formulario:
solid
Ensayo
≥99%
Formulario
solid
densidad
3.339 g/mL at 25 °C (lit.)
cadena SMILES
[Na+].[O-]Br(=O)=O
InChI
1S/BrHO3.Na/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1
Clave InChI
XUXNAKZDHHEHPC-UHFFFAOYSA-M
Aplicación
Useful oxidant for the transformation of tetrahydropyranyl ethers to carbonyl compounds.[1]
Pictogramas
Flame over circleSkull and crossbones
Palabra de señalización
Danger
Frases de peligro
Clasificaciones de peligro
Acute Tox. 3 Oral - Eye Irrit. 2 - Ox. Sol. 1 - Skin Irrit. 2 - STOT SE 3
Órganos de actuación
Respiratory system
Código de clase de almacenamiento
5.1A - Strongly oxidizing hazardous materials
Clase de riesgo para el agua (WGK)
WGK 3
Punto de inflamabilidad (°F)
Not applicable
Punto de inflamabilidad (°C)
Not applicable
Equipo de protección personal
dust mask type N95 (US), Eyeshields, Gloves, type P3 (EN 143) respirator cartridges
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Mohammadpoor-Baltork, I. Nourozi, A.R.
Synthesis, 487-487 (1999)
Wagner A Alves et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 58(7), 1395-1399 (2002-06-27)
The splitting of v1 (794 cm(-1)) and v3 (805 cm(-1)) modes in the stretching region of the bromate (C3v) was observed for the first time in dimethyl sulfoxide (DMSO) using vibrational spectroscopy. Depolarization measurements allowed to assign the asymmetric and
Shu-Ichi Shinohara et al.
Chemical communications (Cambridge, England), (39)(39), 4735-4737 (2008-10-03)
We demonstrate the chemical and optical control of the self-sustaining peristaltic motion of a structural colored porous hydrogel.
Diana I Roncaglia et al.
Physical chemistry chemical physics : PCCP, 13(16), 7426-7432 (2011-03-25)
The Belousov-Zhabotinsky reaction was studied under the influence of nanometric confinements induced by a complex polymer, the PAMAM-G4 dendrimers. They are well-defined in both molecular weight and architecture and are capable of molecular inclusion, making "unimolecular active micelles". The effect
T Umemura et al.
Journal of cancer research and clinical oncology, 119(8), 463-469 (1993-01-01)
Accumulation of alpha 2u-globulin and induction of cell proliferation were examined in kidneys of rats exposed to KBrO3, KBr or NaBrO3 in their drinking water. Hyaline droplets observed after KBrO3 or NaBrO3 administration to male rats were specifically immunostained for