MilliporeSigma
147222
Sigma-Aldrich
Tetrahidrofuran
≥99.5%, contains 250 ppm BHT as inhibitor
Sinónimos:
Oxolano, Óxido de butileno, Óxido de tetrametileno
About This Item
Fórmula empírica (notación de Hill):
C4H8O
Número de CAS:
Peso molecular:
72.11
Beilstein/REAXYS Number:
102391
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
product name
Tetrahidrofuran, ≥99.5%
vapor density
2.5 (vs air)
vapor pressure
114 mmHg ( 15 °C)
143 mmHg ( 20 °C)
assay
≥99.5%
form
liquid
autoignition temp.
610 °F
contains
250 ppm BHT as inhibitor
expl. lim.
1.8-11.8 %
impurities
<0.030% water
refractive index
n20/D 1.407 (lit.)
bp
65-67 °C (lit.)
65-67 °C
mp
−108 °C (lit.)
solubility
water: soluble
density
0.889 g/mL at 25 °C (lit.)
SMILES string
C1CCOC1
InChI
1S/C4H8O/c1-2-4-5-3-1/h1-4H2
InChI key
WYURNTSHIVDZCO-UHFFFAOYSA-N
Legal Information
DuPont product
Optional
Referencia del producto
Descripción
Precios
signalword
Danger
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
target_organs
Central nervous system, Respiratory system
supp_hazards
Storage Class
3 - Flammable liquids
wgk_germany
WGK 1
flash_point_f
-6.2 °F - closed cup
flash_point_c
-21.2 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Todd J Harris et al.
Biomaterials, 31(5), 998-1006 (2009-10-24)
The use of biomaterials for gene delivery can potentially avoid many of the safety concerns with viral gene delivery. However, the efficacy of polymeric gene delivery methods is low, particularly in vivo. One significant concern is that the interior and
Synthetic routes to tetrahydrofuran, tetrahydropyran, and spiroketal units of polyether antibiotics and a survey of spiroketals of other natural products.
Boivin TLB.
Tetrahedron, 43(15), 3309-3362 (1987)
Eagleson M.
Concise Encyclopedia Chemistry, 883-883 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 152-152 (1994)
Polyhedral clathrate hydrates. X. Structure of the double hydrate of tetrahydrofuran and hydrogen sulfide.
Mak TCW, et al.
J. Chem. Phys. , 42(8), 2732-2737 (1965)