Saltar al contenido
MilliporeSigma
Todas las fotos(1)

Key Documents

P5-36-XT-ENCL-DUCT

Air Science USA LLC

Air Science Ducted Fume Hood

Fume Hood 36" Wide

Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización


About This Item

PDF Icon
Shipped & Sold By Air Science
A Trusted Marketplace Seller
PDF Icon
Have a question?

General description

Ducted Fume Hood 36" Wide

Ducted Fume Hoods and Enclosures exhuast harmful fume and particulates away from the operators's breathing zone when connected to an inhouse vent system.


Air Science® Purair® Basic Ducted Fume Hoods are designed to protect personnel from exposure to hazardous agents and toxic chemicals in various applications. These hoods are available in several standard sizes and are extremely compact and lightweight. They are built for simplicity and feature a hinged folding front sash. Clear back, sides, and front panels allow ambient light into the chamber and provide users with an unobstructed view of contents. An open base allows easy placement over a variety of equipment or sinks. Optional worksurfaces are also available.


Specifications:

  • Nominal Width (feet): 3ft
  • Depth: Standard Depth
  • Height: Extra Tall Height
  • Pack Size: Each
  • UOM: EA

  • Design Features:

  • Hinged Front Sash: When closed, the cabinet sash protects the contents from inadvertent external contact and better isolates the air within. The sash is easy to open and close.
  • Steel Support Frame: The chemical-resistant epoxy coated steel frame adds mechanical strength.
  • Optional Work Surface: The internal work surface can be fitted with an optional polypropylene tray.
  • Pass Through Ports: Convenient rear wall passthrough ports for safe routing of instrument cords and cables.

  • Additional Features:

    • 360 Degree Visibility: Clear back and side panels allow ambient light into the chamber and provide users with an unobstructed view of contents.

    Options and Accessories:

  • Airflow Meter: DWYER Airflow Meter.
  • Spill Tray: Black Polypropylene worksurface with spill lip.

  • IMPORTANT NOTE:

    • Important Note: Ducting / fan not included with unit. End user is responsible for meeting exhaust ventilation regulations before venting fumes and should defer to the appropriate agency or agencies (federal, state, local) and / or contact a certified industrial hygienist or qualified safety professional to verify application suitability with this product..

    Legal Information

    Purair is a registered trademark of Air Science USA LLC

    Certificados de análisis (COA)

    Busque Certificados de análisis (COA) introduciendo el número de lote del producto. Los números de lote se encuentran en la etiqueta del producto después de las palabras «Lot» o «Batch»

    ¿Ya tiene este producto?

    Encuentre la documentación para los productos que ha comprado recientemente en la Biblioteca de documentos.

    Visite la Librería de documentos