AATB-67008
AAT Bioquest
AAT Bioquest SYBR Dye qPCR Calibration Plate
1 Plate Optimized for ABI7500 Fast 96-Well
About This Item
fluorescence
λex 498 nm;λem 522 nm
manufacturer/tradename
AAT Bioquest
General description
SYBR Dye qPCR Calibration Plate can be used to maintain your 7500 Real-Time PCR system with Fast 96-well block. This calibration plate is ready to use without any additional preparation steps required.
Legal Information
SYBR is a trademark of Molecular Probes, Inc.