AATB-17700
AAT Bioquest
AAT Bioquest Gelite™ Safe DNA Gel Stain
100 ul 10,000X Water Solution
About This Item
fluorescence
λex 513 nm;λem 552 nm
manufacturer/tradename
AAT Bioquest
General description
Gelite™ Safe has been developed specifically to be less hazardous than EtBr for staining DNA in agarose and acrylamide gels with much higher sensitivity. Gelite™ Safe has greatly improved safety and uncompromised sensitivity. The exceptional sensitivity and strong DNA binding affinity of Gelite™ Safe allows DNA to be stained prior to or post electrophoresis without destaining. Gelite™ Safe was optimized to be compatible with various instruments, including UV and blue-light transiluminators, gel documentation systems, and laser scanners. It is the first single formulation that can be used in either the green or red channel at your preference.
Legal Information
Gelite™ is a trademark of AAT Bioquest