MilliporeSigma
HomeADC ManufacturingMobius® ADC Reactor Single-Use Components Chemical Compatibility
Mobius® ADC Reactor Single-Use Components Chemical Compatibility
Three Mobius Single-Use Bioreactors (10 L, 100 L, 500 L) aligned next to each other. Each bioreactor consists of a cylindrical vessel with multiple ports and tubes attached, mounted on a wheeled support structure for mobility.
Antibody-drug conjugates consist of a highly cytotoxic and potent active pharmaceutical ingredient (API) payload linked to a monoclonal antibody (mAb) capable of targeting a surface antigen on cancer cells. Conjugation of the mAb to the small molecule via the linker requires safe handling and containment due to the toxic nature of the payload. When used for this conjugation step, the single-use Mobius® ADC Reactor not only increases operator safety and process efficiency, scalability, reproducibility, and flexibility, but also decreases the risk of contamination.
The conjugation step typically includes the use of organic solvents, and as such, it is important to evaluate compatibility with the reactor’s single-use components. The studies described below evaluated the physical compatibility of single-use components in the Mobius® ADC Reactor and the level of extractables when exposed to dimethyl sulfoxide (DMSO) and dimethyl acetamide (DMAc).
Single-use Components Evaluated for Physical Compatibility and Extractables
All single-use components were evaluated for extractables and physical compatibility following exposure to 100% or 20% DMSO, 100% or 20% DMAc, and Milli-Q® water. Typical solvent concentrations of DMSO and DMAc typically do not exceed 20% during conjugation. The various resins used in the molded plastic components of the ADC Reactor were evaluated as tensile bars (Table 1).
ItemDescriptionMaterialModel Solvents
1mAb Addition PortPolyethylene100% DMAc, 100% DMSO, and Milli-Q® water
2Impeller CupPolyethylene100% DMAc, 100% DMSO, and Milli-Q® water
3TC PortPolyethylene100% DMAc, 100% DMSO, and Milli-Q® water
4ImpellerPolypropylene100% DMAc, 100% DMSO, and Milli-Q® water
5Port PlatePolyolefin100% DMAc, 100% DMSO, and Milli-Q® water
6TC CapPolypropylene100% DMAc, 100% DMSO, and Milli-Q® water
7EJ Female Luer Barbed FittingPolypropylene100% DMAc, 100% DMSO, and Milli-Q® water
  Pre-Treatment
Gamma Irradiation Dose40 – 65 kGy
Pre-flushNo pre-flushing prior to extraction          
Time between Gamma and Extraction< 8 weeks
Extraction Conditions
Extraction Solvents100% DMAc, 100% DMSO, and Milli-Q® water
Surface Area to Volume Ratio1:1 cm2 / mL
Temperature40 oC
Duration             72 hours
ConditionsOrbital rotation (50 rpm)             
Table 1.Molded plastic components evaluated for extractables and compatibility as tensile bars made from the corresponding resins.
Single-use gaskets and tubing were also evaluated (Table 2). 
ItemDescriptionMaterialModel Solvents
8GasketPlatinum Cured Silicone100% DMAc, 100% DMSO, and Milli-Q® water
9Dip TubeNon-Print PharMed Tubing100% DMAc, 100% DMSO, and Milli-Q® water
10Pharma 50 TubingPlatinum Cured Silicone100% DMAc, 100% DMSO, and Milli-Q® water
11Pharma 65 TubingPlatinum Cured Silicone100% DMAc, 20% DMAc, 100% DMSO, 20% DMSO and Milli-Q® water
12Pharma 80 TubingPlatinum Cured Silicone100% DMAc, 100% DMSO, and Milli-Q® water
13Ultimus® FilmLDPE (Outer Layer) ULDPE (Inner Layer) EVOH (Gas Barrier) ULDPE (Inner Layer) Woven Nylon and EVA (Strength Layer) ULDPE (Fluid Contact Layer)100% DMAC, 20% DMAc, 100% DMSO, 20% DMSO, and Milli-Q® water
Pre-Treatment
Gamma Irradiation Dose40 – 65 kGy
Pre-flushNo pre-flushing prior to extraction          
Time between Gamma and Extraction< 8 weeks
Extraction Conditions
Extraction Solvents100% or 20% DMAc, 100% or 20% DMSO, and Milli-Q® water
Surface Area to Volume RatioVaries depending on tubing ID.  6 : 1 or 2 : 1 cm2 / mL for Ultimus® Film
Temperature40 oC
Duration             72 hours (24 hours for 100% DMAc and 100% DMSO of Ultimus)
ConditionsOrbital rotation (50 rpm)             
Table 2.Gaskets, tubing, and single-use film evaluated for extractables and compatibility.
Experimental Design
Tensile Bars and Tubing Compatibility
Type 1 tensile bars representing the materials used in molded components, along with 6” (15 cm) samples of tubing and 5” x 1” (12 x 2 cm) of the Ultimus® film, were evaluated for tensile strength. Each sample was secured in pneumatic tensile grips and stretched using an Instron machine at a speed of 2” (5 cm) per minute. The percent relative difference between control (gamma irradiated, without solvent exposure) and stretched materials was plotted for each material.
Silicone Gasket Compressibility 
A dynamic mechanical analyzer (DMA) instrument was used to assess the compressibility of post-gamma irradiated gasket samples under different solvent extraction conditions. During testing, the gasket underwent a 2% compression relative to its original thickness; the force required to achieve this deformation was recorded and plotted. 
Extractables Testing 
Analysis of volatile and semi-volatile organic compounds in the 100% DMAc and 100% DMSO extracts was performed using direct inject gas chromatography-mass spectrometry (DI-GC/MS). DI-GC/MS analysis was not performed on the water, 20% DMAc, and 20% DMSO extracts.
Reversed phase HPLC (RP-HPLC) was used to detect non-volatile organic substances in the extraction solution. Absorbance at UV 214 nm was used for detection of extractables.
Physical Compatibility and Extractables Results
Tensile Bars and Tubing Compatibility
None of the post-gamma irradiated tensile bars (Figure 1), tubing material (Figure 2), or Ultimus® film (Figure 3) samples displayed a notable decrease in measured tensile strength compared to the respective controls. These results demonstrated compatibility of the materials with Milli-Q® water, 100% DMAC, and 100% DMSO solvents under extraction conditions (to the exception of Ultimus® film, which was tested with 20% DMSO / DMAC).
None of the post-gamma irradiated tensile bars displayed a notable decrease in measured tensile strength compared to the respective controls for mAb addition ports.
Figure 1a.Relative change in tensile strength of resins used in mAb addition ports following solvent exposure.
None of the post-gamma irradiated tensile bars displayed a notable decrease in measured tensile strength compared to the respective controls for impeller cup.
Figure 1b.Relative change in tensile strength of resins used in impeller cup following solvent exposure.
None of the post-gamma irradiated tensile bars displayed a notable decrease in measured tensile strength compared to the respective controls for TC port.
Figure 1c.Relative change in tensile strength of resins used in TC port following solvent exposure.
None of the post-gamma irradiated tensile bars displayed a notable decrease in measured tensile strength compared to the respective controls for impeller.
Figure 1d.Relative change in tensile strength of resins used in impeller following solvent exposure.
None of the post-gamma irradiated tensile bars displayed a notable decrease in measured tensile strength compared to the respective controls for port plate.
Figure 1e.Relative change in tensile strength of resins used in port plate following solvent exposure.
None of the post-gamma irradiated tensile bars displayed a notable decrease in measured tensile strength compared to the respective controls for TC cap.
Figure 1f.Relative change in tensile strength of resins used in TC cap following solvent exposure.
None of the post-gamma irradiated tensile bars displayed a notable decrease in measured tensile strength compared to the respective controls for EJ female luer barbed fitting.
Figure 1g. Relative change in tensile strength of resins used in EJ female luer barbed fitting following solvent exposure.
None of the post-gamma irradiated tubing material displayed a notable decrease in measured tensile strength compared to the respective controls for dip tube.
Figure 2a.Relative change in tensile strength of dip tube following solvent exposure.
None of the post-gamma irradiated pharma 50 tubing material displayed a notable decrease in measured tensile strength compared to the respective controls for.
Figure 2b.Relative change in tensile strength of pharma 50 tubing following solvent exposure.
None of the post-gamma irradiated pharma 65 material displayed a notable decrease in measured tensile strength compared to the respective controls for pharma 65 tubing.
Figure 2c.Relative change in tensile strength of pharma 65 tubing following solvent exposure.
None of the post-gamma irradiated pharma 80 tubing material displayed a notable decrease in measured tensile strength compared to the respective controls.
Figure 2d.Relative change in tensile strength of pharma 80 tubing following solvent exposure.
None of the post-gamma irradiated Ultimus® film samples displayed a notable decrease in measured tensile strength compared to the respective controls.
Figure 3.Relative change in tensile strength of Ultimus® film following solvent exposure.
As shown in Figure 4, none of the post-gamma irradiated gasket samples displayed a notable decrease in compression force compared to their respective controls. These results demonstrated compatibility of the gasket samples with Milli-Q® water, 100% DMAC, and 100% DMSO solvents under extraction conditions.
None of the post-gamma irradiated gasket samples displayed a notable decrease in compression force compared to their respective controls.
Figure 4.Relative change in compressibility of gasket samples following solvent exposure.
Extractables
The following tables summarize results of the extractables studies. As shown by RP-HPLC and DI-GC/MS, the resins had excellent resistance when exposed to 100% DMAc and DMSO at 40°C for up to 72 hours with no observed visual or functional defects (Table 3). Similar results were obtained for gaskets, tubing, and Ultimus® Film with RP-HPLC (Table 4) and DI-GC/MS (Table 5).
Conjugation being followed by further purification steps such as tangential flow filtration and typically a buffer exchange, additional reduction of leachable components is expected.
Item
Material
HPLC Results in µg /cm2
DI-GC/MS Results in µg /cm2
100% DMAc
100% DMSO
100% DMAc
100% DMSO
Reporting Limit
100 µg /cm2
0.5 µg /cm2
1
mAb Addition Port
Non-Detected
Non-Detected
1,3-Di-tert-butylbenzene (1.8), Alkanes (1.0-2.4)
1,3-Di-tert-butylbenzene (0.9), Alkanes (0.6-0.9)
2
Impeller Cup
Non-Detected
Non-Detected
3
TC Port
Alkanes (0.6-1.5)
Alkanes (0.7)
4
Impeller
Unknown (0.8)
Non-Detected
5
Port Plate
1,3-Di-tert-butylbenzene (0.6)
1,3-Di-tert-butylbenzene (0.7)
6
TC Cap
Non-Detected
Non-Detected
7
EJ Female Luer Barbed Fitting
Non-Detected
Non-Detected
Table 3.Detection of extractables from resins exposed to DMAc and DMSO using RP-HPLC and DI-GC/MS.
Item
Material
DMAc (%)
Reporting Limit
HPLC Results
DMSO (%)
Reporting Limit
HPLC Results
8
Gasket
100
1000 µg /device
Non-Detected
100
1000 µg /device
Non-Detected
9
Dip Tube
100
70 µg /cm2
Non-Detected
100
70 µg /cm2
Non-Detected
10
Pharma 50 Tubing
100
16 µg /cm2
Non-Detected
100
16 µg /cm2
Non-Detected
11
Pharma 65 Tubing
100
7.5 µg /cm2
Non-Detected
100
7.5 µg /cm2
Non-Detected
12
Pharma 80 Tubing
100
48 µg /cm2
Non-Detected
100
48 µg /cm2
Non-Detected
13
Ultimus® Film
100
50 µg /cm2
Non-Detected
100
50 µg /cm2
Non-Detected
20
3.3 µg /cm2
Non-Detected
20
3.3 µg /cm2
Non-Detected
Table 4.Detection of extractables from gaskets, tubing, and Ultimus® film exposed to DMAc and DMSO using RP-HPLC.
Item
Material
DMAc (%)
RL
DI-GC/MS Results
Data Unit
DMSO (%)
RL
DI-GC/MS Results
Data Unit
8
Gasket
100
5
1,3-Di-tert-butylbenzene (21), Bis(2-ethylhexyl) adipate (13), siloxanes (7-82)
µg /device
100
5
1,3-Di-tert-butylbenzene (15), Dimethyl phthalate (7), Bis(2-ethylhexyl) adipate (12)
µg /device
9
Dip Tube*
100
33
Non-Detected
µg /cm2
100
0.4
2,4-Di-tert-butylphenol (0.5), unknown (0.4)
µg /cm2
10
Pharma 50 Tubing
100
0.08
Siloxanes (0.09-103)
µg /cm2
100
0.08
Siloxanes (0.08-4.7)
µg /cm2
11
Pharma 65 Tubing
100
0.04
Mainly siloxanes (0.04-30)
µg /cm2
100
0.04
Mainly siloxanes (0.04-2.1)
µg /cm2
12
Pharma 80 Tubing
100
0.2
Mainly siloxanes (0.2-355)
µg /cm2
100
0.2
Mainly siloxanes (0.3-18)
µg /cm2
13
Ultimus® Film
100
0.3
Mainly alkanes and alkenes (0.4-0.8)
µg /cm2
100
0.3
Non-Detected
µg /cm2
Table 5.Detection of extractables from gaskets and tubing exposed for DMAc and DMSO using DI-GC/MS. *Mineral oil was found in 100% DMAc extracts.
Conclusion
Single-use technologies offer many benefits for production of ADCs including increased operator safety, process efficiency, scalability, reproducibility, and flexibility. As with all single-use systems, understanding solvent compatibility is essential for a successful and safe implementation. As demonstrated by the results described above, the molded plastic components, tubing, gaskets, and Ultimus® film used in the Mobius® ADC Reactor are all compatible with concentrations of DMSO and DMAc that exceed typical conjugation conditions.
If you have questions about our ADC reactor offering, our experts can help you find the right product for your process.