Enzymatic Assay of 5’-Nucleotidase
1. Objective
To standardize a procedure for determining the enzymatic activity of 5’-Nucleotidase.
2. Scope
This procedure applies to all products that have a specification for 5’-nucleotidase. This procedure is NOT to be used to assay 5’-nucleotidase from bovine liver (N2779), or 5’-nucleotidase from Crotalus adamanteus, insoluble or 5’-nucleotidase as an impurity.
3. Definitions
3.1 Purified Water = water from a deionizing system, resistivity > or = 18 MΩ•cm @ 25 ºC
3.2 Unit Definition = One unit will hydrolyze 1.0 μmol of inorganic phosphorus from adenosine 5’-monophosphate per minute at pH 9.0 at 37 ºC.
3.3 STD = Phosphorus Standard
3.4 5’-AMP = Adenosine 5’-Monophosphate
3.5 Pi = Inorganic Phosphate
3.6 TSCR = Taussky-Shorr Color Reagent
4. Discussion
5’-AMP + H2O 5'- Nucleotidase > Adenosine + Pi
5. Responsibilities
Analytical Services laboratory personnel should follow this procedure as written.
6. Safety
Refer to Safety Data Sheets (SDS) for hazards and appropriate handling precautions.
7. Procedure
7.1 CONDITIONS:
T = 37 °C, pH = 9, A660nm, Light path = 1 cm
7.2 METHOD:
Spectrophotometric Stop Reaction
7.3 REAGENTS:
7.3.1 200 mM Glyicine Buffer, pH 9.0 at 37 ºC (Buffer) Prepare in purified water at 15 mg/mL using Glycine (G7126). Adjust to pH 9.0 at 37 ºC with 1 M NaOH.
7.3.2 66 mM Adenosine 5’-Monophosphate Solution (5'-AMP)
7.3.2.1 Prepare in purified water at 22.9 mg/mL using Adenosine 5’-Monphosphate, Sodium Salt, from Yeast (A1752).
7.3.2.2 Correct the Adenosine 5’-Monophosphate concentration for percent water, percent sodium, and percent purity by high pressure liquid chromatography.
7.3.3 200 mM Magnesium Sulfate Solution (MgSO4) Prepare in purified water at 49.3 mg/mL using magnesium sulfate, heptahydrate (M1880).
7.3.4 Phosphorus Standard (STD) Use Phosphorus Standard Solution, 0.645 μ moles/ mL (P3869).
7.3.5 10 N Sulfuric Acid Solution (H2SO4) Prepare in cold purified water at 0.278 mL/mL using sulfuric acid, 95 –98%, A.C.S. Reagent (258105).
7.3.6 10% (w/v) Ammonium Molybdate Solution [(NH4)6MO7O24]
7.3.6.1 Prepare in Reagent 7.3.5 (H2SO4) at 100 mg/mL using ammonium molybdate, tetrahydrate (A7302). This may require more than 8 hours of stirring for dissolution.
7.3.6.2 If Sigma-Aldrich Product No. A7302 is not available, Ammonium Molybdate, Tetrahydrate (M0878) may be used as a replacement.
7.3.6.3 The 10%(w/v) Ammonium Molybdate Solution is stable in the dark for 6 months at room temperature.
7.3.7 Taussky-Shorr Color Reagent (TSCR)
7.3.7.1 Prepare 100 mL by adding 10mLs of Reagent 7.3.6 [(NH4)6MO7O24] to 70 mL of purified water.
7.3.7.2 Then add 5 g of Ferrous Sulfate, Heptahydrate (F7002).
7.3.7.3 Mix until dissolved and dilute to a final volume of 100 mL with purified water.
7.3.8 5’-Nucleotidase Solution (Enzyme)
7.3.8.1 Immediately before use, prepare a solution containing approximately 100 units/mL in cold purified water and swirl until dissolution.
7.3.8.2 Immediately dilute Reagent 7.3.8.1 to 40 – 60 units/mL in cold purified water. Make a fresh dilution for each assay run.
7.4 PROCEDURE
7.4.1 Assay-1
7.4.1.1 Pipette (in milliliters) the following reagents into suitable containers:
Test-1 | Test-2 | Blank-1 | |
---|---|---|---|
Reagent 7.3.1 (Buffer) | 1.50 | 1.50 | 1.50 |
Purified water | 0.24 | 0.24 | 0.24 |
Reagent 7.3.3 (MgSO4) | 0.10 | 0.10 | 0.10 |
Reagent 7.3.2 (5’-AMP) | 0.15 | 0.15 | 0.15 |
7.4.1.2 Mix by swirling and equilibrate to 37 ºC. Then add:
Reagent 7.3.8 (Enzyme) | 0.005 | 0.005 | ----- |
7.4.1.3 Mix by swirling and incubate Test-1 for exactly 1.0 minute at 37 ºC and incubate Test-2 for exactly 2.0 minutes. Then add:
Reagent 7.3.7 (TSCR) | 4.00 | 4.00 | 4.00 |
Reagent 7.3.8 (Enzyme) | ----- | ----- | 0.005 |
Purified Water | 2.00 | 2.00 | 2.00 |
7.4.1.4 Mix by inversion and incubate for five minutes at room temperature. Transfer to suitable cuvettes and measure the A660 nm for Test-1, Test-2, and Blank-1 versus purified water using a suitable spectrophotometer.
7.4.1.5 If results appear inconsistent, then repeat Steps 7.4.1.1 to 7.4.1.4.
7.4.2 Assay-2
7.4.2.1 Pipette (in milliliters) the following reagents into suitable containers:
Test-3 | Test-4 | Blank-2 | |
---|---|---|---|
Reagent 7.3.1 (Buffer) | 1.50 | 1.50 | 1.50 |
Purified water | 0.24 | 0.24 | 0.24 |
Reagent 7.3.3 (MgSO4) | 0.10 | 0.10 | 0.10 |
Reagent 7.3.2 (5’-AMP) | 0.15 | 0.15 | 0.15 |
7.4.2.2 Mix by swirling and equilibrate to 37 ºC. Then add:
Reagent 7.3.8 (Enzyme) | 0.010 | 0.010 | ----- |
7.4.2.3 Mix by swirling and incubate Test-1 for exactly 1.0 minute at 37 ºC and incubate Test-2 for exactly 2.0 minutes. Then add:
Reagent 7.3.7 (TSCR) | 4.00 | 4.00 | 4.00 |
Reagent 7.3.8 (Enzyme) | ----- | ----- | 0.010 |
Purified Water | 2.00 | 2.00 | 2.00 |
7.4.2.4 Mix by inversion and incubate for five minutes at room temperature. Transfer to suitable cuvettes and measure at 660 nm for Test-3, Test-4, and Blank-2 versus purified water using a suitable spectrophotometer.
7.4.2.5 If results appear inconsistent, then repeat Steps 7.4.2.1 to 7.4.2.4.
7.5 STANDARD CURVE
7.5.1 A standard curve is made by pipetting (in milliliters) the following reagents into suitable containers with vented caps:
Std 1 | Std 2 | Std 3 | Std 4 | Std 5 | Std 6 | Std 7 | Std Blank | |
---|---|---|---|---|---|---|---|---|
Reagent 7.3.4 (STD) | 0.05 | 0.20 | 0.40 | 0.60 | 0.80 | 1.00 | 2.00 | ----- |
Purified Water | 3.95 | 3.80 | 3.60 | 3.40 | 3.20 | 3.00 | 2.00 | 4.00 |
Reagent 7.3.7 (TSCR) | 4.00 | 4.00 | 4.00 | 4.00 | 4.00 | 4.00 | 4.00 | 4.00 |
7.5.2 Mix by inversion and incubate for five minutes at room temperature. Transfer to suitable cuvette and measure at A660 nm for standards and standard-blank versus purified water using a suitable spectrophotometer.
7.6 CALCULATIONS
7.6.1 Standard Curve
ΔA660nm Standard = A660nm Std - A660nm Std Blank
Plot the ΔA660nm of the Standards vs μmoles of Phosphorus. Calculate and record the slope, y-intercept, and linear regression(r-square).
7.6.2 Sample Determination
ΔA660nm Sample = A660nmTest-1 or Test-2 - A660nmTest Blank-1
ΔA660nm Sample = A660nm Test-3 or Test-4 - A660nmTest Blank-2
Determine the micromoles of phosphorus liberated using the Standard curve.
Units/mL enzyme = | (µmol of phosphorus released)(df) | |
(0.5)(10)(2.5) |
df = Dilution Factor
0.005= Volume (in milliliter) of enzyme used or
0.010= Volume (in milliliter) of enzyme used
T= Time (in minutes) of assay per unit definition
Units/mg solid = | Units/mL enzyme |
mg solid/mL enzyme |
Units/mg protein = | Units/mL enzyme |
mg protein/mL enzyme |
7.7 FINAL ASSAY CONCENTRATION :
In a 2 mL reaction mix, the final concentrations are 150 mM glycine, 10 mM magnesium sulfate, 5.0 mM adenosine 5’-monophosphate, and 0.2 – 0.6 units of 5’-nucleotidase.
Network error: Failed to fetch
Reference