How to Adjust the Volume on the Transferpette® from BRAND®
This video explains how to adjust the volume on the Transferpette® before being ready to pipette.
The Transferpette® S is the perfect manual pipette for demanding laboratory scientists who prefer a traditional control layout. The Transferpette® S offers robust yet lightweight construction, superior autoclavability, simple, one-handed operation, and high degree of accuracy and precision for long lasting reliability. The single channel model is available in fixed volume and digital models in volume ranges from 0.1 µL to 10 mL. The multichannel model is available in 8- or 12-channels in volume ranges from 0.5 µL to 300 µL.
Network error: Failed to fetch