HPLC Analysis of Flavonoids on Ascentis® RP-Amide
Conditions | |
---|---|
column | Ascentis® RP-Amide, 15 cm x 4.6 mm I.D., 5 µm particles (565324-U) |
column temp. | 30 °C |
mobile phase | [A] 0.085% orthophosphoric acid; [B] acetonitrile; |
flow rate | 1.0 mL/min |
sample | as indicated in mobile phase A |
injection | 10 µL |
detector | UV, 280 nm |
Description | |
---|---|
Categories | Analytical Chromatography, Flavors, Fragrances |
Featured Industry | Vitamins, Nutraceuticals, and Natural Products |
suitability | application for HPLC |
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch