MilliporeSigma
US EPA Method 8015 (modified): GC Analysis of Gasoline Range Organics (GRO) on SLB®-5ms, Direct Injection
US EPA Method 8015 (modified): GC Analysis of Gasoline Range Organics (GRO) on SLB®-5ms, Direct Injection suitable for GC
Materials
Analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.50 μm
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 1.00 μm
SLB®-5ms Capillary GC Column
L × I.D. 20 m × 0.18 mm, df 0.18 μm
Guard column
Product No.
Description
Pricing
Intermediate Polar Fused Silica Tubing
L × I.D. 5.0 m × 0.25 mm
accessory
Product No.
Description
Pricing
Wheaton wide-mouth bottle
amber soda-lime glass bottle, capacity 1000 mL, white polypropylene cap, (PTFE faced LDPE liner), case of 12 ea
Supeltex M-2B Double-Tapered Ferrule
I.D. 0.4 mm, 0.10-0.25 mm to like I.D. Column, pkg of 2 ea
Capillary Column Butt Connector
body only (ferrules not included)
standard
Product No.
Description
Pricing
N-Nitrosodiphenylamine solution
certified reference material, 5000 μg/mL in methanol
Perfluorotributylamine (PFTBA)
analytical standard
EPA 8270 GC-MS Tuning Solution II
certified reference material, 1000 μg/mL each component in dichloromethane
EPA GRO Mix
certified reference material, in methanol (varied)
EPA CLP Semivolatile Calibration Mix
certified reference material, 1000 μg/mL each component in dichloromethane : benzene (3:1)
EPA CLP Semivolatiles Surrogate Standards Mix
certified reference material, 4000 μg/mL each component in methylene chloride
EPA 8270 Semivolatile Internal Standard Mix
certified reference material, TraceCERT®, 2000 μg/mL each component in methylene chloride, ampule of 2 × 1 mL
EPA 8270 Benzidines Mix
certified reference material, TraceCERT®, 2000 μg/mL each component in methanol, ampule of 1 mL
CONDITIONS
column
SLB-5ms, 30 m x 0.25 mm I.D., 1.0 μm (28476-U)
oven
50 °C (3 min), 15 °C/min to 200 °C
inj. temp.
150 °C
detector
FID, 250 °C
carrier gas
helium, 1.3 mL/min., constant
injection
0.5 μL, 50:1 split
liner
2 mm I.D., FocusLiner inlet liner
sample
EPA GRO Mix, 9 analytes at various concentrations in methanol (47577-U)
Description
Legal Information
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany