GC Analysis of Permanent Gases and Light Hydrocarbons on Carboxen®-1006 PLOT
![GC Analysis of Permanent Gases and Light Hydrocarbons on Carboxen®-1006 PLOT suitable for GC](/deepweb/assets/sigmaaldrich/product/images/304/054/7d90dc40-7a42-4cfd-bb7b-207ab65c9390/800/7d90dc40-7a42-4cfd-bb7b-207ab65c9390.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Carboxen-1006 PLOT, 30 m x 0.53 mm I.D. (25461)
oven
35 °C (1.0 min), 24 °C/min to 250 °C
inj. temp.
200 °C
detector
TCD, 230 °C
carrier gas
helium, 10.0 mL/min
injection
10.0 μL, direct using a sample valve, 150 °C
sample
Permanent gas and light hydrocarbon mix, analytes at concentrations indicated
Description
Legal Information
Carboxen is a registered trademark of Merck KGaA, Darmstadt, Germany