MilliporeSigma
UHPLC/MS Analysis of Insulin Analogs on BIOshell A160 Peptide C18
UHPLC/MS Analysis of Insulin Analogs on BIOshell A160 Peptide C18 application for UHPLC-MS
CONDITIONS
column
BIOshell A160 Peptide C18, 15 cm x 2.1 mm I.D., 2.0 μm particles (67243-U)
mobile phase
[A] 75:25 (0.1% DFA in water: 0.1% DFA in acetonitrile); [B] 50:50 (0.1% DFA in water: 0.1% DFA in acetonitrile)
gradient
23% B for 7 min; 23% B to 100% B in 1 min; hold at 100% B for 7 min
flow rate
0.2 mL/min
pressure
2500 psi (172 bar)
column temp.
75 °C
detector
ESI(+), TIC 100-3000 m/z
injection
0.5 μL
sample
Analog Mixture, 20 μg/mL, 6:3:1 methanol:water:acetic acid containing 0.1% rat plasma
Description
Legal Information
null