MilliporeSigma
HPLC Analysis of Proteins on TSKgel® Ether-5PW
HPLC Analysis of Proteins on TSKgel® Ether-5PW application for HPLC
Materials
analytical column
Product No.
Description
Pricing
TSKgel® Ether-5PW HPLC Column
phase ether, L × I.D. 7.5 cm × 7.5 mm, 10 μm particle size
CONDITIONS
column
TSKgel Ether-5PW, 7.5 cm × 7.5 mm I.D., 10 μm particles (808641)
mobile phase
0.1 M phosphate buffer, pH 7.0 plus ammonium sulfate, 1.8 M to 0 M in 60 min
flow rate
1 mL/min
detector
UV, 280 nm
Description
Legal Information
TSKgel is a registered trademark of Tosoh Corporation