MilliporeSigma
HPLC Analysis of Bromacil Enantiomers on Astec® CHIROBIOTIC® T2
HPLC Analysis of Bromacil Enantiomers on Astec® CHIROBIOTIC® T2 application for HPLC
Materials
Standard
Product No.
Description
Pricing
Bromacil
PESTANAL®, analytical standard
CONDITIONS
column
CHIROBIOTIC T2, 25 cm x 4.6 mm I.D., 5 μm particles (16024AST)
mobile phase
[A] water; [B] ethanol; (70:30, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
1 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC