MilliporeSigma
HPLC Analysis of Propranolol Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of Propranolol Enantiomers on Astec® CHIROBIOTIC® T application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® T Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
standard
Product No.
Description
Pricing
(R)-(+)-Propranolol hydrochloride
≥98% (TLC)
(S)-(−)-Propranolol hydrochloride
≥98% (TLC), powder
(±)-Propranolol hydrochloride
≥99% (TLC), powder
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
15 mM ammonium formate in methanol
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 220 nm
injection
5 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC