MilliporeSigma
HPLC Analysis of Steroids on Ascentis® Express F5 with Methanolic Mobile Phase
HPLC Analysis of Steroids on Ascentis® Express F5 with Methanolic Mobile Phase application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
Ascentis® Express F5, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
CONDITIONS
column
Ascentis Express F5, 10 cm x 2.1 mm I.D., 2.7 μm particles (53569-U)
mobile phase
[A] water [B] methanol; (40:60, A:B)
flow rate
0.2 mL/min
column temp.
35 °C
detector
UV, 254 nm
injection
0.2 mL/min
sample
50 mg/L in 75:25, water:methanol
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany