HPLC Analysis of Various Polymers on TSKgel® SuperMultiporeHZ-H
Materials
analytical column
Product No.
Description
Pricing
TSKgel® SuperMultiporeHZ-H HPLC Column
phase polystyrene-divinylbenzene, L × I.D. 15 cm × 4.6 mm, 6 μm particle sizeguard column
Product No.
Description
Pricing
TSKgel® SuperMPHZ-H Guard Column
phase polystyrene-divinylbenzene, L × I.D. 2 cm × 4.6 mm, 6 μm particle sizerelated product
Product No.
Description
Pricing
TSKgel® SuperMultiporeHZ-M HPLC Column
phase polystyrene-divinylbenzene, L × I.D. 15 cm × 4.6 mm, 4 μm particle sizeTSKgel® SuperMultiporeHZ-N HPLC Column
phase polystyrene-divinylbenzene, L × I.D. 15 cm × 4.6 mm, 3 μm particle sizeTSKgel® SuperMPHZ-M Guard Column
phase polystyrene-divinylbenzene, L × I.D. 2 cm × 4.6 mm, 4 μm particle sizeTSKgel® SuperMPHZ-N Guard Column
phase polystyrene-divinylbenzene, L × I.D. 2 cm × 4.6 mm, 3 μm particle sizeCONDITIONS
column
TSKgel SuperMultiporeHZ-N, 15 cm x 4.6 mm I.D., 6 μm particles (821885)
mobile phase
THF
flow rate
0.35 mL/min
column temp.
40 °C
detector
refractive index (RI)
injection
10 μL
sample
each polymer, 3 mg/mL
Description
Other Notes
Data provided by Tosoh Bioscience LLC.
Legal Information
TSKgel is a registered trademark of Tosoh Corporation