MilliporeSigma
HPLC Analysis of Ulcer Treatment Drugs on Ascentis® RP-Amide
HPLC Analysis of Ulcer Treatment Drugs on Ascentis® RP-Amide application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® RP-Amide HPLC Column
5 μm particle size, L × I.D. 15 cm × 2.1 mm
CONDITIONS
column
Ascentis RP-Amide, 15 cm x 2.1 mm I.D., 5 μm particles (565305-U)
mobile phase
[A] 10 mM ammonium phosphate, pH 6.2 with phosphoric acid; [B] acetonitrile
gradient
5 to 15% B in 10 min; to 55% B in 8 min; held for 5 min
flow rate
0.2 mL/min
column temp.
35 °C
detector
UV, 220 nm
injection
5 μL
sample
as indicated in 95:5 water:methanol
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany