HPLC Analysis of Taxols on Discovery® HS F5
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery HS F5, 15 cm × 2.1 mm I.D., 3 μm particles (567503-U)
mobile phase
[A] 10 mM ammonium acetate (pH 6.8 unadjusted); [B] acetonitrile
flow rate
0.2 mL/min
column temp.
ambient
detector
UV, 227 nm
injection
10 μL
sample
10 μg/mL (in 50:50, water:acetonitrile)
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany