MilliporeSigma
HPLC Analysis of Proteins on TSKgel® Butyl-NPR
HPLC Analysis of Proteins on TSKgel® Butyl-NPR application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
TSKgel® Butyl-NPR HPLC Column
phase C4 (butyl), L × I.D. 3.5 cm × 4.6 mm, 2.5 μm particle size
CONDITIONS
column
TSKgel Butyl-NPR, 3.5 cm × 4.6 mm I.D., 2.5 μm particles (814947)
mobile phase
[A] 0.1 M phosphate buffer, pH 7.0 plus ammonium sulfate, 2.3 M to 0 M in 12 min [B & C] = 0.1 M phosphate buffer, pH 7.0 plus ammonium sulfate, 1.8 M to 0 M in 60 min
flow rate
1 mL/min
detector
UV, 280 nm
Description
Legal Information
TSKgel is a registered trademark of Tosoh Corporation