MilliporeSigma
HPLC Analysis of Proteins (2) on BIOshell A400 Protein C4
HPLC Analysis of Proteins (2) on BIOshell A400 Protein C4 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
BIOshell A400 Protein C4, 3.4 μm HPLC Column
3.4 μm particle size, L × I.D. 10 cm × 2.1 mm
CONDITIONS
sample
protein mixture, g/L ea: superoxide dismutase, 0.3; bovine serum albumin, 0.3; alpha-lactalbumin, 0.2; beta-lactoglobulin A, 0.3; catalase, 0.3; (carbonic anhydrase, 0.3; beta-Amylase, 1.0; glucose-6-phosphate dehydrogenase, 1.0; thioredoxin, 0.3; phosphorylase b, 2.5.)
column
BIOshell A400 Protein C4, 10 cm X 2.1 mm I.D., 3.4 μm particles (66825-U)
mobile phase
[A] 75:25, (0.1% TFA in water):(0.1% TFA in acetonitrile) ; [B] 25:75, (0.1% TFA in water):(0.1% TFA in acetonitrile)
gradient
15 to 75% B in 10 min
flow rate
0.3 mL/min
pressure
2410 psi (166 bar)
column temp.
30 °C
detector
UV, 215 nm
injection
1 μL
Description
Analysis Note
This application demonstrates the utility of the BIOshell A400 Protein C4 column for the reversed-phase chromatography of proteins from a variety of sources and of different molecular weights.
Legal Information
null