HPLC Analysis of Indapamide Enantiomers on Astec® Cellulose DMP
![HPLC Analysis of Indapamide Enantiomers on Astec® Cellulose DMP application for HPLC](/deepweb/assets/sigmaaldrich/product/images/267/543/3e6cd707-658b-4a0e-8459-ed436cb0152e/800/3e6cd707-658b-4a0e-8459-ed436cb0152e.jpg)
CONDITIONS
column
Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 μm particles (51098AST)
mobile phase
[A] methanol; [B] DEA; (100:0.1, A:B)
flow rate
0.5 mL/min
column temp.
25 °C
detector
UV, 230 nm
injection
2 μL
sample
2 mg/mL in mobile phase