MilliporeSigma
HPLC Analysis of α-Hydroxy Aliphatic Acids on Discovery® RP-Amide C16
HPLC Analysis of α-Hydroxy Aliphatic Acids on Discovery® RP-Amide C16 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Discovery® RP-AmideC16 HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Discovery RP-AmideC16, 15 cm × 4.6 mm I.D., 5 μm particles (505013)
mobile phase
25 mM potassium phosphate, pH 3.0
flow rate
1 mL/min
column temp.
30 °C
detector
UV, 220 nm
injection
10 μL
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany