HPLC Analysis of Antibiotics on Discovery® C18
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery C18, 15 cm × 4.6 mm I.D., 5 μm particles (504955)
mobile phase
[A] 25 mM potassium phosphate (pH 3.0); [B] acetonitrile
flow rate
1 mL/min
column temp.
35 °C
detector
UV, 220 nm
injection
10 μL
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany