MilliporeSigma
HPLC Analysis of Anti-Inflammatory Drugs on Discovery® RP-Amide C16
HPLC Analysis of Anti-Inflammatory Drugs on Discovery® RP-Amide C16 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Discovery® RP-AmideC16 HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Discovery RP-Amide C16, 15 cm × 4.6 mm I.D., 5 μm particles (505013)
mobile phase
(A) acetonitrile; (B) 25 mM KH2PO4, pH 3.0; (40:60, A:B)
flow rate
1 mL/min
column temp.
30 °C
detector
UV, 230 nm
injection
10 μL, 1 μg/mL each analyte
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany