MilliporeSigma
HPLC Analysis of 3′,4′-Methylenedioxy-α-pyrrolidinopropiophenone (MDPPP) Enantiomers on Astec® CHIROBIOTIC® V2
HPLC Analysis of 3′,4′-Methylenedioxy-α-pyrrolidinopropiophenone (MDPPP) Enantiomers on Astec® CHIROBIOTIC® V2 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® V2 Chiral HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Astec CHIROBIOTIC V2 15 cm x 4.6 mm I.D., 5 μm particles (15023AST)
mobile phase
[A] methanol; [B] water; [C] acetic acid; [D] ammonium hydroxide; (95:5:0.1:0.02, A:B:C:D)
flow rate
1 mL/min
pressure
1100 psi (70 bar)
column temp.
20 °C
detector
UV, 251 nm
injection
5 μL
sample
100 μg/mL in methanol
Description
Analysis Note
3′,4′-Methylenedioxy-a-pyrrolidinopropiophenone, also known as MDPPP, is a synthetic cathinone found in numerous bath salt blends under the street name NRG-1. This designer drug first emerged in Germany in the late 1990s and early 2000s as an ingredient in imitation ecstasy pills. Astec CHIROBIOTIC V2, based on vancomycin CSP, provided resolution of the enatiomers. CRMs from Cerillliant provided reliable identification and quantification.
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC