MilliporeSigma
HPLC Analysis of 4-Methyl-5-Phenyl-2-Oxazolidone Enantiomers on Astec® CHIROBIOTIC® TAG
HPLC Analysis of 4-Methyl-5-Phenyl-2-Oxazolidone Enantiomers on Astec® CHIROBIOTIC® TAG application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® TAG Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
Standard
Product No.
Description
Pricing
(4S,5R)-(−)-4-Methyl-5-phenyl-2-oxazolidinone
99%
(4R,5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinone
99%
CONDITIONS
column
CHIROBIOTIC TAG, 25 cm x 4.6 mm I.D., 5 μm particles (14024AST)
mobile phase
methanol
flow rate
0.8 mL/min
column temp.
23 °C
detector
UV, 230 nm
injection
2 μL
sample
2 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC