MilliporeSigma
HPLC Analysis of Diet Soda Spiked with Neotame on Ascentis® Express RP-Amide
HPLC Analysis of Diet Soda Spiked with Neotame on Ascentis® Express RP-Amide application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
Ascentis® Express RP-Amide, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 3 cm × 4.6 mm
standard
Product No.
Description
Pricing
Caffeine
Sigma Reference Standard, vial of 250 mg
Aspartame
analytical standard
SS Benzoic Acid
2000 μg/mL in dichloromethane, analytical standard
Neotame
analytical standard
CONDITIONS
column
Ascentis Express RP-Amide, 3 cm x 4.6 mm I.D., 2.7 μm particles (53921-U)
mobile phase
[A] 100 mM ammonium acetate, pH 5.6 with acetic acid; [B] water; [C] acetonitrile
gradient
20% A constant; 75 to 20% B, 5 to 60% C in 1 min; held at 20% B, 60% C for 0.1 min
flow rate
3 mL/min
pressure
3336 psi (230 bar)
column temp.
40 °C
detector
UV, 214 nm
injection
1 mL/min
sample
diet soda spiked with neotame 350 μg/mL
Description
Analysis Note
This application demonstrates the suitability of Ascentis Express RP-Amide for the efficient separation of common drink additives used in diet cola.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany