MilliporeSigma
GC Analysis of Pyrazines in Peanut Butter on SUPELCOWAX 10 after SPME using 50/30 μm DVB/Carboxen/PDMS Fiber
GC Analysis of Pyrazines in Peanut Butter on SUPELCOWAX 10 after SPME using 50/30 μm DVB/Carboxen/PDMS Fiber suitable for GC, application for SPME
Materials
Analytical column
Product No.
Description
Pricing
SUPELCOWAX 10 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
Related product
Product No.
Description
Pricing
SPME Fiber Holder
for use with manual sampling
SPME Fiber Holder
for use with CTC CombiPAL and Gerstel MPS2
SPME fiber
Product No.
Description
Pricing
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS)
needle size 24 ga, for use with manual holder
Standard
Product No.
Description
Pricing
Carbon disulfide
anhydrous, ≥99%
2-Ethylpyrazine
≥98%, FG
2-Methylpyrazine
Pharmaceutical Secondary Standard; Certified Reference Material
Dimethyl disulfide
analytical standard
2,3-Diethylpyrazine
analytical reference material
2,5-Dimethylpyrazine
analytical standard
CONDITIONS
sample/matrix
5 g peanut butter in 40 mL vial
SPME fiber
divinylbenzene-Carboxen-polydimethylbenzene StableFlex (57328-U)
extraction
headspace, 30 min at 65 °C in heating block
desorption process
270 °C for 5 min
column
SUPELCOWAX 10, 30 m × 0.25 mm, 0.25 μm (24079)
oven
40 °C (5 min) to 230 °C at 4 °C/min
inj. temp.
270 °C
injection
splitless/split, closed 0.5 min, 270 °C, with 0.75 mm liner
detector
ion trap mass spectrometer, m/z =30-350 at 0.6 sec/scan. Selected ions used for quantitation.
carrier gas
Helium, 30 cm/sec
liner
0.75 mm
sample
peanut butter
Description
Analysis Note
In this application, SPME was used for the analysis of volatiles in multifloral honey using a headspace method. Headspace SPME (HS-SPME) using a Carboxen/PDMS/DVB fiber coupled with gas chromatography-mass spectrometry (GC/MS) was used for the characterization of the volatile fraction of honey samples. Using SPME it was possible to obtain and distinguish the chemical fingerprints of different types of honey. The relatively polar SUPELCOWAX 10 GC column provides excellent separation of these polar compounds.
Legal Information
null