MilliporeSigma
GC Analysis of Lactone Enantiomers on β-DEX 110
GC Analysis of Lactone Enantiomers on β-DEX™ 110 suitable for GC
Materials
analytical column
Product No.
Description
Pricing
β-DEX 110
L × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
β-DEX 110, 30 m × 0.25 mm I.D., 0.25 μm (24301)
oven
90 °C, 1 °C/min to 200 °C
inj. temp.
300 °C
detector
FID, 300 °C
carrier gas
helium, 20 cm/sec @ 90 °C
injection
1 μL, 100:1 split
sample
0.5 mg/mL in methylene chloride
Description
Legal Information
DEX is a trademark of Sigma-Aldrich Co. LLC