MilliporeSigma
GC Analysis of Volatiles in Water on SPB®-1 after SPME using 100 μm PDMS Fiber
GC Analysis of Volatiles in Water on SPB®-1 after SPME using 100 μm PDMS Fiber suitable for GC, application for SPME
Materials
analytical column
Product No.
Description
Pricing
SPB®-1 Capillary GC Column
L × I.D. 10 m × 0.20 mm, df 1.20 μm
SPME fiber
Product No.
Description
Pricing
SPME Fiber Assembly
100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, plain red hub
CONDITIONS
SPME fiber
polydimethylsiloxane, 100 μm (57300-U)
extraction
5 min immersed in water with rapid stirring
column
SPB-1, 10 m × 0.20 mm I.D., 1.2 μm (24134-U)
oven
40 °C ( 0.75 min) to 160 °C at 20 °C/min
inj. temp.
230 °C, splitless (closed 3 min)
detector
FID, 260 °C
carrier gas
helium, 40 cm/sec
sample
50ppb each analyte in 1.8 mL saturated salt water
Description
Legal Information
SPB is a registered trademark of Merck KGaA, Darmstadt, Germany