MilliporeSigma
GC Analysis of Peppermint Essential Oil on SLB®-5ms
GC Analysis of Peppermint Essential Oil on SLB®-5ms suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven
50 °C, 3 °C/min to 300 °C
inj. temp.
220 °C
detector
FID, 300 °C
carrier gas
helium, 30 cm/sec
detector
1 μL, 100:1 split
sample
Peppermint oil purchased from Misitano&Stracuzzi S.p.a. (Messina, Italy) diluted 1:10 in n-hexane
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany