MilliporeSigma
US EPA Method TO-17: GC Analysis of Volatiles on VOCOL® after Collection/Desorption using Air Toxics Tube
US EPA Method TO-17: GC Analysis of Volatiles on VOCOL® after Collection/Desorption using Air Toxics Tube suitable for GC, application for air monitoring
Materials
Analytical column
Product No.
Description
Pricing
VOCOL® Capillary GC Column
L × I.D. 60 m × 0.25 mm, df 1.50 μm
standard
Product No.
Description
Pricing
EPA TO-14 Calibration Mix 1 (39 components)
39 components, 100 ppb each component in nitrogen, pkg of 110 L (pi-marked cylinder), analytical standard
CONDITIONS
sample/matrix
10-50 ng each analyte introduced onto tube (calibration standard, Cat. No. 41900)
adsorbent tube
Air Toxics (glass) (25086)
column
VOCOL, 60 m × 0.25 mm I.D., 1.5 μm film (24154)
oven
37 °C (4 min) to 200 °C at 4 °C/min, hold 10 min
detector
FID
carrier gas
helium, 2 mL/min
Description
Legal Information
VOCOL is a registered trademark of Merck KGaA, Darmstadt, Germany