MilliporeSigma
US EPA Method 610: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SLB®-5ms
US EPA Method 610: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SLB®-5ms suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SPB®-608 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (24103-U)
oven
70 °C (0.2 min), 25 °C/min to 265 °C, 5 °C/min to 315 °C (2 min)
inj. temp.
250 °C
detector
FlD, 350 °C
carrier gas
helium, 1.3 mL/min, constant
injection
2.0 μL pulsed splitless (30 psi until 0.2 min)
liner
4 mm I.D. single taper
sample
1 μg/mL PAH standard in methylene chloride
Description
Analysis Note
US EPA Method 610 describes the analysis of polynuclear aromatic hydrocarbons (commonly referred to as PAHs or PNAs) by both HPLC and GC. The method designates the use of a packed GC column, however capillary columns will provide better resolution, baseline stability, and longer lifetime. This application illustrates the separation of the target compound list in this method on the SLB-5ms capillary GC column. A pressure pulsed injection was used to improve response of the heavier PAHs, and to reduce .the larger vapor volume resulting from a 2 μL injection.
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany