MilliporeSigma
GC Analysis of Short and Long Chain Free Fatty Acids on Nukol
GC Analysis of Short and Long Chain Free Fatty Acids on Nukol suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Nukol Capillary GC Column
size × I.D. 15 m × 0.53 mm, df 0.50 μm
CONDITIONS
column
Nukol, 15 m x 0.53 mm I.D., 0.50 μm (25326)
oven
100 °C, 10 °C/min to 220 °C
detector
FID
carrier gas
helium, 30 mL/min
injection
0.5 μL, direct injection
sample
16 analytes, at various concentrations from 50 to 800 μg/mL
Description
Legal Information
null