MilliporeSigma
GC Analysis of Distilled Lime Essential Oil on Equity-1 and Equity-5
GC Analysis of Distilled Lime Essential Oil on Equity-1 and Equity-5 suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Equity-1 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
Equity-1, 30 m × 0.25 mm I.D., 0.25 μm (28046-U)
column
Equity-5, 30 m × 0.25 mm I.D., 0.25 μm (28089-U)
oven
75 °C (8 min), 4 °C/min to 200 °C (10 min)
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 cm/sec @ 110 °C
injection
Wet Needle, 100:1 split
liner
split, cup design
sample
distilled lime oil
Description
Legal Information
null