MilliporeSigma
GC Analysis of Class 2 Residual Solvents on Equity-5
GC Analysis of Class 2 Residual Solvents on Equity-5 suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Equity-5 Capillary GC Column
L × I.D. 30 m × 0.53 mm, df 5.00 μm
guard column
Product No.
Description
Pricing
Intermediate Polar Fused Silica Tubing
L × I.D. 5.0 m × 0.53 mm
CONDITIONS
column
Equity-5, 30 m x 0.53 mm I.D., 5.0 μm (28279-U) with 5 m intermediate polarity guard (25339) (28279-U)
oven
35 °C (15 min), 5 °C/min to 200 °C
inj. temp.
225 °C
detector
FID, 250 °C
carrier gas
helium, 30 cm/sec @ 35 °C
injection
1 μL, 33:1 split
liner
single taper
sample
28 analytes, at concentrations indicated in DMSO
Description
Legal Information
null