Merck
GC Analysis of Aroma Fusel Alcohols in Tennessee Whiskey on Watercol 1910
GC Analysis of Aroma Fusel Alcohols in Tennessee Whiskey on Watercol™ 1910 suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Watercol 1910 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.20 μm
CONDITIONS
column
Watercol 1910, 30 m x 0.25 mm I.D., 0.20 μm; two identical columns installed in the same inlet, each going to a separate detector (29711-U)
oven
50 °C (1 min), 5 °C/min to 180 °C (3 min)
inj. temp.
250 °C
detector
FID, 200 °; TCD, 200 °C
carrier gas
helium, 25 cm/sec (measured in each column at an oven temperature of 125 °C)
injection
1 μL, 100:1 split
liner
4 mm I.D., split/splitless type, wool packed single taper FocusLiner design (2879925-U)
sample
neat Tennessee whiskey
Description
Analysis Note
Fusel alcohols are by-products of the fermentation process. Four of these (n-propanol, isobutanol, active amyl alcohol, and isoamyl alcohol) are important to alcoholic beverage manufacturers due to their aroma characteristics.
The "Watercol" series of ionic liquid capillary GC columns are characterized by their ability to produce a sharp peak shape for water, sharp enough so that water can be integrated and quantified. Sharp peak shapes are also produced for many other small polar analytes, such as the four aroma fusel alcohols.
In this application, a Tennessee whiskey sample was analyzed for the presence of aroma fusel alcohols. As shown, great peak shapes were obtained, resulting in good resolution. In addition to sharp fusel alcohol peaks, the column′s selectivity results in elution of both ethanol and water free from all analytes.
Legal Information
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
Watercol is a trademark of Sigma-Aldrich Co. LLC