MilliporeSigma
  • Novel mutations in the 3 region of the polycystic kidney disease 1 (PKD1) gene.
Novel mutations in the 3 region of the polycystic kidney disease 1 (PKD1) gene.
Human genetics (2000-01-27)
A R Afzal, M Hand, E Ternes-Pereira, A Saggar-Malik, R Taylor, S Jeffery
ABSTRACT
Mutation screening in 90 unrelated ADPKD1 patients was carried out on some of the exons in the single copy area (37, 38, 39, 44, 45) using genomic PCR and SSCP. Four novel mutations were found: a 15 bp in-frame deletion in exon 39 [nt11449 (del 15)], a 2 bp deletion in exon 44 [nt12252 (del 2)], a G insertion in exon 44 [nt12290 (Ins G)], and a GTT in-frame deletion in exon 45 [nt12601 (del 3)].