MilliporeSigma
  • Lack of CD27-CD45RA-V gamma 9V delta 2+ T cell effectors in immunocompromised hosts and during active pulmonary tuberculosis.
Lack of CD27-CD45RA-V gamma 9V delta 2+ T cell effectors in immunocompromised hosts and during active pulmonary tuberculosis.
Journal of immunology (Baltimore, Md. : 1950) (2002-01-22)
Cristiana Gioia, Chiara Agrati, Rita Casetti, Cristiana Cairo, Giovanna Borsellino, Luca Battistini, Giorgio Mancino, Delia Goletti, Vittorio Colizzi, Leopoldo P Pucillo, Fabrizio Poccia
ABSTRACT
In humans, the circulating pool of mycobacteria-reactive Vgamma9Vdelta2+ T cells is expanded with age and may contribute to Mycobacterium tuberculosis immunosurveillance. We observed that two subsets of Vgamma9Vdelta2+ T cells could be identified on the basis of CD27 expression in immunocompetent adults, showing that functionally differentiated gammadelta T cells have lost CD27 expression. In contrast, the CD27-CD45RA-Vgamma9Vdelta2+ T cell subset of effector cells was absent in cord blood cells from healthy newborns and lacking in the peripheral blood from HIV-infected patients. Moreover, circulating Vgamma9Vdelta2+ T cell effectors were significantly reduced in patients with acute pulmonary tuberculosis, resulting in a reduced frequency of IFN-gamma-producing cells after stimulation with nonpeptidic mycobacterial ligands. These observations indicate that monitoring and boosting gammadelta T cell effectors could be clinically relevant both in immunocompromised hosts and during active tuberculosis disease.