MilliporeSigma
  • Autoregulation of human relaxin-2 gene expression critically involves relaxin and glucocorticoid receptor binding to glucocorticoid response half-sites in the relaxin-2 promoter.
Autoregulation of human relaxin-2 gene expression critically involves relaxin and glucocorticoid receptor binding to glucocorticoid response half-sites in the relaxin-2 promoter.
Regulatory peptides (2009-03-18)
Thomas Dschietzig, Cornelia Bartsch, Silja Wessler, Gert Baumann, Karl Stangl
ABSTRACT
Relaxin peptides act in brain, reproductive and cardiovascular systems, kidneys, and connective tissue through different G protein-coupled receptors. We reported that human relaxin-2 and porcine relaxin are both agonists at the human glucocorticoid receptor (GR). Here, we investigated the possible auto-regulation of relaxin-2 gene expression via recently discovered GR-binding sites in the relaxin-2 promoter. We found that porcine relaxin increased the secretion of human relaxin-like immunoreactivity in HeLa and THP-1 cells. Silencing of GR gene expression completely abolished this effect whereas transfection of wild-type GR into naturally GR-devoid HT-29 cells established relaxin sensitivity. Relaxin was shown to stimulate CAT expression driven by different deletion constructs of the 5'-flanking region of the relaxin-2 promoter. In chromatin immunoprecipitation assays, we detected both GR and relaxin binding to the relaxin-2 promoter. Gel shift assays indicated binding of relaxin-activated GR to half-GREs located between 160 and 200 bp upstream of transcription start but not to the GRE at -900 bp. Relaxin bound to human GR and displaced established GR agonists. Immunofluorescence experiments visualized nuclear co-localization of relaxin and GR in response to relaxin. In conclusion, we have identified a positive auto-regulatory loop of human relaxin-2 expression which involves GR and relaxin/GR binding to half-GREs in the relaxin-2 promoter.